siRNA Details
Gene Symbol: Vps4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145678.1 | NP_663711.1 | 6 | 560 | ||
XM_006255560.1 | XP_006255622.1 | 6 | 779 | |||
GenBank mRNA | AB076398.1 | - | 6 | 560 | ||
BC070931.1 | - | 6 | 663 | 7096867 | ||
FQ213218.1 | - | 740 |
