siRNA Details
| Gene Symbol: Ptpn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145683.1 | NP_663716.1 | 4 | 469 | ||
| XM_006249820.2 | XP_006249882.1 | 4 | 944 | |||
| XM_006249821.2 | XP_006249883.1 | 4 | 599 | |||
| GenBank mRNA | U28356.1 | - | 4 | 469 | ||

Ptpn7
Genomic Map