siRNA Details
| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022669.1 | NP_073160.1 | 1 | 1093 | ||
| XM_008767195.1 | XP_008765417.1 | 2 | 1417 | |||
| GenBank mRNA | M93669.1 | - | 1 | 1093 | ||
| X13618.1 | - | 1 | 1093 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scg2| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022669.1 | NP_073160.1 | 1 | 1093 | ||
| XM_008767195.1 | XP_008765417.1 | 2 | 1417 | |||
| GenBank mRNA | M93669.1 | - | 1 | 1093 | ||
| X13618.1 | - | 1 | 1093 | |||