siRNA Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012648.1 | NP_036780.1 | 13 | 1715 | ||
| XM_008759682.1 | XP_008757904.1 | 13 | 1750 | |||
| XM_008759683.1 | XP_008757905.1 | 14 | 1960 | |||
| XM_008759684.1 | XP_008757906.1 | 13 | 1792 | |||
| GenBank mRNA | EU627516.1 | - | 12 | 1660 | ||
| U35174.1 | - | 12 | 1670 | |||
| U35175.1 | - | 12 | 1670 | |||
| X77932.1 | - | 13 | 1732 | |||

Scnn1b
Genomic Map