siRNA Details
| Gene Symbol: Ccl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031530.1 | NP_113718.1 | 3 | 504 | ||
| GenBank mRNA | AF058786.1 | - | 3 | 504 | ||
| M57441.1 | - | 3 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl2| Gene Symbol: Ccl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031530.1 | NP_113718.1 | 3 | 504 | ||
| GenBank mRNA | AF058786.1 | - | 3 | 504 | ||
| M57441.1 | - | 3 | 468 | |||