siRNA Details
| Gene Symbol: Slc9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113335.1 | NP_001106806.1 | 2 | 721 | ||
| NM_012653.2 | NP_036785.2 | 1 | 718 | |||
| GenBank mRNA | L11004.1 | - | 1 | 733 | ||
| L11236.1 | - | 2 | 721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a2| Gene Symbol: Slc9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113335.1 | NP_001106806.1 | 2 | 721 | ||
| NM_012653.2 | NP_036785.2 | 1 | 718 | |||
| GenBank mRNA | L11004.1 | - | 1 | 733 | ||
| L11236.1 | - | 2 | 721 | |||