siRNA Details
| Gene Symbol: Syp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012664.2 | NP_036796.1 | 4 | 803 | ||
| GenBank mRNA | BC099798.1 | - | 818 | 7936715 | ||
| X06177.1 | - | 812 | ||||
| X06388.1 | - | 844 | ||||
| X06655.1 | - | 776 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syp| Gene Symbol: Syp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012664.2 | NP_036796.1 | 4 | 803 | ||
| GenBank mRNA | BC099798.1 | - | 818 | 7936715 | ||
| X06177.1 | - | 812 | ||||
| X06388.1 | - | 844 | ||||
| X06655.1 | - | 776 | ||||