siRNA Details
| Gene Symbol: Tacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012667.2 | NP_036799.1 | 1440 | |||
| GenBank mRNA | J05097.1 | - | 1440 | |||
| M31477.1 | - | 4 | 897 | |||
More Information
Other Species Targeted
Set Membership: |
|

Tacr1
Genomic Map