siRNA Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012668.2 | NP_036800.1 | 12 | 1395 | ||
| GenBank mRNA | BC089813.1 | - | 12 | 1395 | 7372826 | |
| M18340.1 | - | 12 | 1448 | |||
| X02741.1 | - | 12 | 1448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tat| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012668.2 | NP_036800.1 | 12 | 1395 | ||
| GenBank mRNA | BC089813.1 | - | 12 | 1395 | 7372826 | |
| M18340.1 | - | 12 | 1448 | |||
| X02741.1 | - | 12 | 1448 | |||