siRNA Details
| Gene Symbol: Tg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270783.1 | NP_001257712.1 | 37 | 6473 | ||
| NM_030988.2 | NP_112250.2 | 38 | 6617 | |||
| GenBank mRNA | AB035201.1 | - | 38 | 6617 | ||
| AB679745.1 | - | 39 | 6591 | |||
| AB682738.1 | - | 38 | 6591 | |||
| AB682739.1 | - | 38 | 6447 | |||
| AF221622.1 | - | 38 | 6591 | |||
| X02318.1 | - | 11 | 1174 | |||

Tg
Genomic Map