siRNA Details
Gene Symbol: Tg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270783.1 | NP_001257712.1 | 46, 47 | 8075 | ||
NM_030988.2 | NP_112250.2 | 47, 48 | 8219 | |||
GenBank mRNA | AB035201.1 | - | 47, 48 | 8219 | ||
AB679745.1 | - | 48, 49 | 8193 | |||
AB682738.1 | - | 47, 48 | 8193 | |||
AB682739.1 | - | 47, 48 | 8049 | |||
AF221622.1 | - | 47, 48 | 8193 | |||
X02318.1 | - | 20, 21 | 2776 |
