siRNA Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270854.1 | NP_001257783.1 | 5 | 521 | ||
| NM_012672.3 | NP_036804.2 | 5 | 521 | |||
| XM_008770500.1 | XP_008768722.1 | 3 | 433 | |||
| GenBank mRNA | DQ191165.1 | - | 2 | 268 | ||
| J03819.1 | - | 5 | 517 | |||
| J03933.1 | - | 5 | 518 | |||

Thrb
Genomic Map