siRNA Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270854.1 | NP_001257783.1 | 11 | 1490 | ||
NM_012672.3 | NP_036804.2 | 12 | 1598 | |||
XM_006251709.2 | XP_006251771.1 | 7 | 1511 | |||
XM_008770500.1 | XP_008768722.1 | 9 | 1402 | |||
GenBank mRNA | DQ191165.1 | - | 9 | 1345 | ||
HM043807.1 | - | 8 | 1387 | |||
J03819.1 | - | 11 | 1486 | |||
J03933.1 | - | 11 | 1487 |
