siRNA Details
| Gene Symbol: Tp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030989.3 | NP_112251.2 | 8 | 1103 | ||
| XM_006246594.2 | XP_006246656.1 | 9 | 1532 | |||
| XM_006246595.2 | XP_006246657.1 | 8 | 1889 | |||
| XM_008767773.1 | XP_008765995.1 | 8 | 1110 | |||
| GenBank mRNA | BC081788.1 | - | 8 | 1032 | 7193583 | |
| BC098663.1 | - | 8 | 1061 | 7384467 | ||
| FQ232659.1 | - | 8 | 1091 | |||
| U90328.1 | - | 7 | 972 | |||
| X13058.1 | - | 7 | 995 | |||

Tp53
Genomic Map