siRNA Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100634.2 | NP_001094104.1 | 12 | 1389 | ||
| XM_006229218.2 | XP_006229280.1 | 11 | 1515 | |||
| XM_006229219.1 | XP_006229281.1 | 12 | 1428 | |||
| GenBank mRNA | CB609064.1 | - | 5 | 504 | ||
| CB764690.1 | - | 1 | 35 | |||
| X53501.1 | - | 12 | 1389 | |||

Tph1
Genomic Map