siRNA Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100634.2 | NP_001094104.1 | 7 | 711 | ||
| XM_006229218.2 | XP_006229280.1 | 6 | 837 | |||
| XM_006229219.1 | XP_006229281.1 | 7 | 750 | |||
| GenBank mRNA | CB796568.1 | - | 2 | 173 | ||
| CB809951.1 | - | 3 | 274 | |||
| M28000.1 | - | 1 | 93 | |||
| X53501.1 | - | 7 | 711 | |||

Tph1
Genomic Map