siRNA Details
| Gene Symbol: Pgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133284.2 | NP_579818.1 | 4 | 489 | ||
| GenBank mRNA | X04644.1 | - | 4 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgc| Gene Symbol: Pgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133284.2 | NP_579818.1 | 4 | 489 | ||
| GenBank mRNA | X04644.1 | - | 4 | 489 | ||