siRNA Details
| Gene Symbol: Vhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052801.1 | NP_434688.1 | 2 | 426 | ||
| GenBank mRNA | S80345.1 | - | 2 | 367 | ||
| U14746.1 | - | 2 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vhl| Gene Symbol: Vhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052801.1 | NP_434688.1 | 2 | 426 | ||
| GenBank mRNA | S80345.1 | - | 2 | 367 | ||
| U14746.1 | - | 2 | 426 | |||