siRNA Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031534.2 | NP_113722.2 | 6 | 830 | ||
| XM_006234620.1 | XP_006234682.1 | 5 | 1172 | |||
| XM_006234621.2 | XP_006234683.1 | 6 | 1301 | |||
| XM_006234622.2 | XP_006234684.1 | 5 | 1250 | |||
| GenBank mRNA | X69716.1 | - | 6 | 830 | ||

Wt1
Genomic Map