siRNA Details
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031534.2 | NP_113722.2 | 3, 4 | 675 | ||
XM_006234620.1 | XP_006234682.1 | 3, 4 | 1068 | |||
XM_006234621.2 | XP_006234683.1 | 3, 4 | 1146 | |||
XM_006234622.2 | XP_006234684.1 | 3, 4 | 1146 | |||
GenBank mRNA | X69716.1 | - | 3, 4 | 675 |
