siRNA Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012689.1 | NP_036821.1 | 6 | 1433 | ||
| XM_006227832.2 | XP_006227894.1 | 6 | 2033 | |||
| XM_008758703.1 | XP_008756925.1 | 7 | 2081 | |||
| XM_008758704.1 | XP_008756926.1 | 6 | 2005 | |||
| GenBank mRNA | AB477039.1 | - | 6 | 1439 | ||
| GU111761.1 | - | 2 | 190 | |||
| KJ746832.1 | - | 5 | 1224 | |||
| KJ746833.1 | - | 5 | 1224 | |||
| X61098.1 | - | 5 | 1297 | |||
| Y00102.1 | - | 6 | 1433 | |||

Esr1
Genomic Map