siRNA Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017061.2 | NP_058757.1 | 5 | 1124 | ||
| XM_006254713.2 | XP_006254775.1 | 3 | 1207 | |||
| XM_006254714.2 | XP_006254776.1 | 4 | 1107 | |||
| XM_006254715.2 | XP_006254777.1 | 3 | 1208 | |||
| GenBank mRNA | BC078861.1 | - | 5 | 1124 | 7113042 | |
| FQ230374.1 | - | 3 | 359 | |||
| FQ231737.1 | - | 2 | 148 | |||
| U11038.1 | - | 3 | 1092 | |||

Lox
Genomic Map