siRNA Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017061.2 | NP_058757.1 | 6, 7 | 1346 | ||
| XM_006254713.2 | XP_006254775.1 | 4, 5 | 1429 | |||
| XM_006254714.2 | XP_006254776.1 | 5, 6 | 1329 | |||
| XM_006254715.2 | XP_006254777.1 | 4, 5 | 1430 | |||
| GenBank mRNA | BC078861.1 | - | 6, 7 | 1346 | 7113042 | |
| FQ230374.1 | - | 4, 5 | 581 | |||
| FQ231737.1 | - | 3, 4 | 370 | |||
| U11038.1 | - | 4, 5 | 1314 | |||

Lox
Genomic Map