siRNA Details
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017065.1 | NP_058761.1 | 8 | 1254 | ||
GenBank mRNA | AY742860.1 | - | 1584 | |||
X15468.1 | - | 8 | 1254 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017065.1 | NP_058761.1 | 8 | 1254 | ||
GenBank mRNA | AY742860.1 | - | 1584 | |||
X15468.1 | - | 8 | 1254 |