siRNA Details
| Gene Symbol: Cd8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031538.2 | NP_113726.1 | 1 | 27 | ||
| XM_006236630.2 | XP_006236692.1 | 2 | 656 | |||
| XM_008762976.1 | XP_008761198.1 | 2 | 404 | |||
| GenBank mRNA | BC088126.1 | - | 1 | 27 | 7381324 | |
| X03015.1 | - | 1 | 66 | |||

Cd8a
Genomic Map