siRNA Details
| Gene Symbol: Grpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012706.2 | NP_036838.1 | 2 | 662 | ||
| GenBank mRNA | X56661.1 | - | 2 | 754 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grpr| Gene Symbol: Grpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012706.2 | NP_036838.1 | 2 | 662 | ||
| GenBank mRNA | X56661.1 | - | 2 | 754 | ||
Set Membership: |
|