siRNA Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172091.2 | NP_742088.1 | 7 | 665 | ||
| NM_172092.2 | NP_742089.1 | 7 | 673 | |||
| XM_006247876.2 | XP_006247938.1 | 7 | 779 | |||
| XM_006247877.1 | XP_006247939.1 | 5 | 504 | |||
| XM_008768468.1 | XP_008766690.1 | 6 | 722 | |||
| XM_008768469.1 | XP_008766691.1 | 6 | 725 | |||
| GenBank mRNA | BC081700.1 | - | 6 | 530 | 7192550 | |
| CV117246.1 | - | 3 | 189 | |||
| L04796.1 | - | 7 | 635 | |||
| M96674.1 | - | 7 | 675 | |||
| X68692.1 | - | 7 | 635 | |||
More Information
Set Membership: |
|

Gcgr
Genomic Map