siRNA Details
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017072.1 | NP_058768.1 | 5 | 498 | ||
GenBank mRNA | M11710.1 | - | 5 | 637 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017072.1 | NP_058768.1 | 5 | 498 | ||
GenBank mRNA | M11710.1 | - | 5 | 637 |