siRNA Details
| Gene Symbol: Pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017328.2 | NP_059024.1 | 1 | 393 | ||
| GenBank mRNA | FQ214696.1 | - | 1 | 406 | ||
| FQ214983.1 | - | 1 | 406 | |||
| FQ215677.1 | - | 1 | 406 | |||
| FQ217291.1 | - | 1 | 413 | |||
| FQ218013.1 | - | 1 | 406 | |||
| FQ223775.1 | - | 1 | 406 | |||
| FQ224280.1 | - | 1 | 406 | |||
| FQ224543.1 | - | 1 | 409 | |||
| M31835.1 | - | 1 | 394 | |||

Pgam2
Genomic Map