siRNA Details
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212546.4 | NP_997711.1 | 2 | 385 | ||
GenBank mRNA | BC108294.1 | - | 1 | 247 | 7111250 | |
BC108297.1 | - | 2 | 385 | 7115041 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212546.4 | NP_997711.1 | 2 | 385 | ||
GenBank mRNA | BC108294.1 | - | 1 | 247 | 7111250 | |
BC108297.1 | - | 2 | 385 | 7115041 |