siRNA Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270575.1 | NP_001257504.1 | 501 | |||
| NM_001270576.1 | NP_001257505.1 | 413 | ||||
| NM_030991.3 | NP_112253.1 | 5 | 501 | |||
| GenBank mRNA | AB003991.1 | - | 292 | |||
| AB003992.1 | - | 4 | 292 | |||
| AF245227.1 | - | 5 | 500 | |||
| BC087699.1 | - | 5 | 456 | 7314318 | ||
| DY315301.1 | - | 433 | ||||
| FM033263.1 | - | 413 | ||||
| FQ212040.1 | - | 5 | 472 | |||
| FQ213289.1 | - | 6 | 469 | |||
| FQ213923.1 | - | 5 | 472 | |||
| FQ214162.1 | - | 5 | 501 | |||
| U56261.1 | - | 265 | ||||
| U56262.1 | - | 4 | 265 | |||

Snap25
Genomic Map