siRNA Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270575.1 | NP_001257504.1 | 5, 6 | 614 | ||
| NM_001270576.1 | NP_001257505.1 | 5, 6 | 526 | |||
| NM_030991.3 | NP_112253.1 | 5, 6 | 614 | |||
| GenBank mRNA | AB003991.1 | - | 405 | |||
| AB003992.1 | - | 4, 5 | 405 | |||
| AF245227.1 | - | 5, 6 | 613 | |||
| BC087699.1 | - | 5, 6 | 569 | 7314318 | ||
| DY315301.1 | - | 5, 6 | 546 | |||
| FM033263.1 | - | 5, 6 | 526 | |||
| FQ212040.1 | - | 5, 6 | 585 | |||
| FQ213923.1 | - | 5, 6 | 585 | |||
| FQ214162.1 | - | 5, 6 | 614 | |||
| U56261.1 | - | 378 | ||||
| U56262.1 | - | 4, 5 | 378 | |||

Snap25
Genomic Map