siRNA Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172305.1 | NP_001165776.1 | 16, 17 | 1861 | ||
| NM_012713.3 | NP_036845.3 | 16, 17 | 1861 | |||
| GenBank mRNA | K03486.1 | - | 6, 7 | 624 | ||
| M13706.1 | - | 14, 15 | 1871 | |||
| M15522.1 | - | 4, 5 | 390 | |||
| M19007.1 | - | 14, 15 | 1821 | |||
| X04139.1 | - | 4, 5 | 390 | |||
| X04440.1 | - | 14, 15 | 2415 | |||

Prkcb
Genomic Map