siRNA Details
Gene Symbol: Ampd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138876.1 | NP_620231.1 | 3 | 145 | ||
XM_006233060.2 | XP_006233122.1 | 2 | 222 | |||
XM_008761369.1 | XP_008759591.1 | 2 | 145 | |||
GenBank mRNA | J02811.1 | - | 3 | 145 |
