siRNA Details
| Gene Symbol: Pigr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012723.4 | NP_036855.3 | 266 | |||
| GenBank mRNA | FQ104960.1 | - | 3 | 266 | ||
| X15741.1 | - | 2 | 160 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigr| Gene Symbol: Pigr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012723.4 | NP_036855.3 | 266 | |||
| GenBank mRNA | FQ104960.1 | - | 3 | 266 | ||
| X15741.1 | - | 2 | 160 | |||