siRNA Details
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012727.3 | NP_036859.2 | 12 | 1181 | ||
XM_006254523.2 | XP_006254585.1 | 11 | 1123 | |||
GenBank mRNA | BC128706.1 | - | 12 | 1181 | 7455982 | |
J04446.1 | - | 2 | 256 | |||
J04600.1 | - | 208 | ||||
M63334.1 | - | 1075 | ||||
M64757.1 | - | 10 | 1171 | |||
S65840.1 | - | 11 | 1106 |
