siRNA Details
| Gene Symbol: Glp1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012728.1 | NP_036860.1 | 10 | 1003 | ||
| GenBank mRNA | M97797.1 | - | 10 | 1003 | ||
| S75952.1 | - | 10 | 997 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glp1r| Gene Symbol: Glp1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012728.1 | NP_036860.1 | 10 | 1003 | ||
| GenBank mRNA | M97797.1 | - | 10 | 1003 | ||
| S75952.1 | - | 10 | 997 | |||
Set Membership: |
|