siRNA Details
| Gene Symbol: Cyp2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012730.1 | NP_036862.1 | 4 | 658 | ||
| GenBank mRNA | AB008423.1 | - | 4 | 534 | ||
| BC078897.1 | - | 4 | 593 | 7109496 | ||
| FQ218496.1 | - | 4 | 599 | |||
| M16655.1 | - | 4 | 584 | |||
| M22330.1 | - | 4 | 658 | |||

Cyp2d2
Genomic Map