siRNA Details
Gene Symbol: Slc12a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270617.1 | NP_001257546.1 | 14 | 1937 | ||
NM_001270618.1 | NP_001257547.1 | 14 | 1937 | |||
XM_008762140.1 | XP_008760362.1 | 16 | 2201 | |||
XM_008762141.1 | XP_008760363.1 | 16 | 2200 | |||
XM_008762142.1 | XP_008760364.1 | 15 | 2105 | |||
XM_008762143.1 | XP_008760365.1 | 15 | 2105 | |||
XM_008762144.1 | XP_008760366.1 | 14 | 2009 | |||
GenBank mRNA | BC087017.1 | - | 14 | 1979 | 7109809 | |
EF577032.1 | - | 13 | 1921 | |||
JN579708.1 | - | 14 | 1937 | |||
U10096.1 | - | 14 | 1937 |
