siRNA Details
| Gene Symbol: Scarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031541.1 | NP_113729.1 | 11 | 1482 | ||
| GenBank mRNA | AB002151.1 | - | 11 | 1482 | ||
| AF071495.1 | - | 11 | 1289 | |||
| AY451993.1 | - | 11 | 1387 | |||
| AY682847.1 | - | 11 | 1387 | |||
| BC076504.1 | - | 11 | 1391 | 7099863 | ||
| D89655.1 | - | 11 | 1400 | |||
| U76205.1 | - | 11 | 1328 | |||

Scarb1
Genomic Map