siRNA Details
| Gene Symbol: Pld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030992.1 | NP_112254.1 | 5, 6 | 723 | ||
| XM_006232205.2 | XP_006232267.1 | 5, 6 | 1498 | |||
| XM_006232206.2 | XP_006232268.1 | 6, 7 | 1151 | |||
| XM_008760891.1 | XP_008759113.1 | 6, 7 | 1151 | |||
| GenBank mRNA | AB000778.1 | - | 5, 6 | 738 | ||
| AB000779.1 | - | 5, 6 | 723 | |||
| AB003170.1 | - | 5, 6 | 615 | |||
| AB003171.1 | - | 5, 6 | 615 | |||
| AF017251.1 | - | 4, 5 | 567 | |||
| U69550.1 | - | 5, 6 | 723 | |||
| U88986.1 | - | 5, 6 | 635 | |||

Pld1
Genomic Map