siRNA Details
| Gene Symbol: Chrna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052805.2 | NP_434692.2 | 5 | 1241 | ||
| GenBank mRNA | AY574254.1 | - | 5 | 1241 | ||
| L31621.1 | - | 5 | 1317 | |||
| X03440.1 | - | 5 | 1391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna3| Gene Symbol: Chrna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052805.2 | NP_434692.2 | 5 | 1241 | ||
| GenBank mRNA | AY574254.1 | - | 5 | 1241 | ||
| L31621.1 | - | 5 | 1317 | |||
| X03440.1 | - | 5 | 1391 | |||