siRNA Details
| Gene Symbol: Hsd11b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017081.2 | NP_058777.1 | 4 | 838 | ||
| XM_006255445.2 | XP_006255507.1 | 4 | 1554 | |||
| GenBank mRNA | BC087023.1 | - | 4 | 825 | 7130250 | |
| U22424.1 | - | 4 | 839 | |||

Hsd11b2
Genomic Map