siRNA Details
| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012746.1 | NP_036878.1 | 11 | 1649 | ||
| GenBank mRNA | M76706.1 | - | 11 | 1649 | ||
| M83746.1 | - | 11 | 1624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk2| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012746.1 | NP_036878.1 | 11 | 1649 | ||
| GenBank mRNA | M76706.1 | - | 11 | 1649 | ||
| M83746.1 | - | 11 | 1624 | |||