siRNA Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034164.1 | NP_001029336.1 | 16 | 1624 | ||
| NM_032612.3 | NP_116001.2 | 16 | 1624 | |||
| GenBank mRNA | AF205604.1 | - | 14 | 1334 | ||
| BC062079.1 | - | 16 | 1624 | 5599645 | ||
| FQ227362.1 | - | 1 | 43 | |||
| FQ231334.1 | - | 1 | 44 | |||

Stat1
Genomic Map