siRNA Details
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017084.1 | NP_058780.1 | 3 | 396 | ||
| GenBank mRNA | X06150.1 | - | 3 | 396 | ||
| Gene Symbol: LOC100911564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766920.1 | XP_008765142.1 | 3 | 428 | ||

Gnmt
Genomic Map