siRNA Details
| Gene Symbol: Gfra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012750.1 | NP_036882.1 | 4 | 624 | ||
| XM_006252238.2 | XP_006252300.1 | 3 | 1393 | |||
| GenBank mRNA | AF003825.1 | - | 1174 | |||
| AF005226.1 | - | 4 | 504 | |||
| AY059644.1 | - | 2 | 99 | |||
| AY059645.1 | - | 3 | 174 | |||
| U97143.1 | - | 4 | 624 | |||

Gfra2
Genomic Map