siRNA Details
| Gene Symbol: Fyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012755.1 | NP_036887.1 | 11 | 1316 | ||
| XM_006256520.2 | XP_006256582.1 | 9 | 2151 | |||
| XM_006256521.2 | XP_006256583.1 | 9 | 2142 | |||
| XM_006256522.1 | XP_006256584.1 | 10 | 1603 | |||
| GenBank mRNA | FQ231621.1 | - | 2 | 201 | ||
| FQ232093.1 | - | 4 | 416 | |||
| U35365.1 | - | 11 | 1316 | |||

Fyn
Genomic Map