siRNA Details
| Gene Symbol: Sp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012761.1 | NP_036893.1 | 3 | 1205 | ||
| GenBank mRNA | U07610.1 | - | 3 | 1205 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001166385, NM_009239, U62522, AB221633, BC076630.

Sp4
Genomic Map