siRNA Details
| Gene Symbol: Sp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012761.1 | NP_036893.1 | 3 | 1093 | ||
| GenBank mRNA | U07610.1 | - | 3 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp4| Gene Symbol: Sp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012761.1 | NP_036893.1 | 3 | 1093 | ||
| GenBank mRNA | U07610.1 | - | 3 | 1093 | ||